Assay Details
| Gene Symbol: Ppara | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013072444.2 | XP_012927898.1 | 4 - 5 | 1151 | 59 | |
| XM_013072445.2 | XP_012927899.1 | 4 - 5 | 1151 | 59 | ||
| XM_021258801.1 | XP_021114460.1 | 5 - 6 | 837 | 59 | ||
| XM_021258802.1 | XP_021114461.1 | 5 - 6 | 846 | 59 | ||
| XM_021258803.1 | XP_021114462.1 | 4 - 5 | 1151 | 59 | ||
More Information
Set Membership: |
|

Ppara
Genomic Map