Assay Details
Gene Symbol: Fmc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013074494.2 | XP_012929948.1 | 1 - 2 | 999 | 68 | |
XM_021263688.1 | XP_021119347.1 | 1 - 2 | 1012 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
