Assay Details
| Gene Symbol: Umad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013078892.2 | XP_012934346.1 | 3 - 4 | 329 | 103 | |
| XM_021242860.1 | XP_021098519.1 | 4 - 5 | 313 | 103 | ||
| XM_021242861.1 | XP_021098520.1 | 3 - 4 | 245 | 103 | ||
More Information
Set Membership: |
|

Umad1
Genomic Map