Assay Details
Gene Symbol: Pmepa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004840931.2 | XP_004840988.1 | 2 - 3 | 288 | 108 | |
XM_021255158.1 | XP_021110817.1 | 2 - 3 | 224 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() |
