Assay Details
| Gene Symbol: Tcfl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004841026.2 | XP_004841083.1 | 1 - 2 | 599 | 102 | |
| XM_004841027.2 | XP_004841084.1 | 1 - 2 | 599 | 102 | ||
| XM_021255228.1 | XP_021110887.1 | 1 - 2 | 599 | 102 | ||
More Information
Set Membership: |
|

Tcfl5
Genomic Map