Assay Details
Gene Symbol: LOC101705246 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004843107.3 | XP_004843164.2 | 1 - 2 | 756 | 116 | |
XM_013071584.2 | XP_012927038.1 | 1 - 2 | 806 | 116 | ||
XM_021256942.1 | XP_021112601.1 | 1 - 2 | 237 | 116 | ||
XM_021256946.1 | XP_021112605.1 | 1 - 2 | 807 | 116 | ||
XM_021256948.1 | XP_021112607.1 | 1 - 2 | 803 | 116 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
