Assay Details
Gene Symbol: Diablo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004843888.3 | XP_004843945.1 | 5 - 6 | 1229 | 69 | |
XM_004843891.3 | XP_004843948.1 | 5 - 6 | 606 | 69 | ||
XM_021257554.1 | XP_021113213.1 | 5 - 6 | 1274 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
