Assay Details
Gene Symbol: Washc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004844847.2 | XP_004844904.1 | 5 - 6 | 524 | 103 | |
XM_004844848.2 | XP_004844905.1 | 5 - 6 | 497 | 103 | ||
XM_004844849.2 | XP_004844906.1 | 4 - 5 | 378 | 103 | ||
XM_021258409.1 | XP_021114068.1 | 4 - 5 | 434 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
