Assay Details
Gene Symbol: LOC101708403 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004845613.3 | XP_004845670.1 | 1 - 2 | 129 | 75 | |
XM_004845614.3 | XP_004845671.1 | 1 - 2 | 132 | 75 | ||
XM_013072565.2 | XP_012928019.1 | 1 - 2 | 132 | 75 | ||
XM_013072566.2 | XP_012928020.1 | 1 - 2 | 129 | 75 | ||
XM_021258996.1 | XP_021114655.1 | 1 - 2 | 132 | 75 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004845613.3, XM_004845614.3, XM_013072565.2, XM_013072566.2, XM_021258996.1 transcripts.
Set Membership: |
![]() ![]() |
Panther Classification:
