Assay Details
Gene Symbol: LOC101708403 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004845613.3 | XP_004845670.1 | 5 - 6 | 701 | 94 | |
XM_004845614.3 | XP_004845671.1 | 5 - 6 | 704 | 94 | ||
XM_013072565.2 | XP_012928019.1 | 5 - 6 | 704 | 94 | ||
XM_013072566.2 | XP_012928020.1 | 5 - 6 | 701 | 94 | ||
XM_021258995.1 | XP_021114654.1 | 5 - 6 | 670 | 94 | ||
XM_021258996.1 | XP_021114655.1 | 5 - 6 | 704 | 94 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
