Assay Details
Gene Symbol: LOC101708403 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004845613.3 | XP_004845670.1 | 9 - 10 | 1355 | 62 | |
XM_013072565.2 | XP_012928019.1 | 10 - 11 | 1419 | 62 | ||
XM_021258995.1 | XP_021114654.1 | 10 - 11 | 1385 | 62 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
