Assay Details
| Gene Symbol: Tnip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004847383.2 | XP_004847440.1 | 4 - 5 | 942 | 59 | |
| XM_021260393.1 | XP_021116052.1 | 4 - 5 | 946 | 59 | ||
More Information
Set Membership: |
|

Tnip2
Genomic Map