Assay Details
Gene Symbol: LOC101697371 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004851668.3 | XP_004851725.1 | 10 - 11 | 1326 | 75 | |
XM_021264027.1 | XP_021119686.1 | 7 - 8 | 1502 | 75 | ||
XM_021264029.1 | XP_021119688.1 | 9 - 10 | 1309 | 75 | ||
XM_021264030.1 | XP_021119689.1 | 7 - 8 | 1255 | 75 | ||
XM_021264031.1 | XP_021119690.1 | 9 - 10 | 1159 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
