Assay Details
Gene Symbol: LOC101697371 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004851668.3 | XP_004851725.1 | 5 - 6 | 755 | 87 | |
XM_021264026.1 | XP_021119685.1 | 5 - 6 | 729 | 87 | ||
XM_021264027.1 | XP_021119686.1 | 4 - 5 | 738 | 87 | ||
XM_021264029.1 | XP_021119688.1 | 4 - 5 | 738 | 87 | ||
XM_021264030.1 | XP_021119689.1 | 4 - 5 | 738 | 87 | ||
XM_021264031.1 | XP_021119690.1 | 5 - 6 | 755 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() |
