Assay Details
Gene Symbol: Efcc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004855640.2 | XP_004855697.2 | 1 - 2 | 1345 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
