Assay Details
Gene Symbol: LOC101705956 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004856680.2 | XP_004856737.1 | 2 - 3 | 691 | 94 | |
XM_021237823.1 | XP_021093482.1 | 2 - 3 | 470 | 94 | ||
XM_021237824.1 | XP_021093483.1 | 2 - 3 | 696 | 94 | ||
XM_021237825.1 | XP_021093484.1 | 2 - 3 | 579 | 94 | ||
XM_021237826.1 | XP_021093485.1 | 2 - 3 | 470 | 94 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
