Assay Details
Gene Symbol: Pdzd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004856783.3 | XP_004856840.1 | 9 - 10 | 1617 | 82 | |
XM_004856784.3 | XP_004856841.1 | 9 - 10 | 1614 | 82 | ||
XM_021237724.1 | XP_021093383.1 | 9 - 10 | 1800 | 82 | ||
XM_021237725.1 | XP_021093384.1 | 9 - 10 | 1797 | 82 | ||
XM_021237726.1 | XP_021093385.1 | 7 - 8 | 2318 | 82 | ||
XM_021237727.1 | XP_021093386.1 | 8 - 9 | 2181 | 82 | ||
XM_021237728.1 | XP_021093387.1 | 8 - 9 | 1591 | 82 | ||
XM_021237729.1 | XP_021093388.1 | 8 - 9 | 1587 | 82 | ||
XM_021237730.1 | XP_021093389.1 | 6 - 7 | 2188 | 82 | ||
XM_021237731.1 | XP_021093390.1 | 6 - 7 | 1337 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
