Assay Details
Gene Symbol: Rab40b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004861046.2 | XP_004861103.1 | 3 - 4 | 401 | 67 | |
XM_013078206.2 | XP_012933660.1 | 3 - 4 | 355 | 67 | ||
XM_021241119.1 | XP_021096778.1 | 2 - 3 | 1185 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
