Assay Details
| Gene Symbol: Cbarp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004865974.3 | XP_004866031.1 | 5 - 6 | 665 | 81 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cbarp| Gene Symbol: Cbarp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004865974.3 | XP_004866031.1 | 5 - 6 | 665 | 81 | |
Set Membership: |
|