Assay Details
| Gene Symbol: Pcdhac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004921122.2 | XP_004921179.1 | 1 | 2523 | 88 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pcdhac1| Gene Symbol: Pcdhac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004921122.2 | XP_004921179.1 | 1 | 2523 | 88 | |
Set Membership: |
|