Assay Details
Gene Symbol: Ten1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021241065.1 | XP_021096724.1 | 3 - 4 | 434 | 78 | |
XM_021241066.1 | XP_021096725.1 | 4 - 5 | 540 | 78 | ||
XM_021241067.1 | XP_021096726.1 | 3 - 4 | 313 | 78 | ||
XM_021241068.1 | XP_021096727.1 | 3 - 4 | 314 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
