Assay Details
Gene Symbol: Aste1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021242549.1 | XP_021098208.1 | 4 - 5 | 1792 | 56 | |
XM_021242596.1 | XP_021098255.1 | 4 - 5 | 1932 | 56 | ||
XM_021242629.1 | XP_021098288.1 | 4 - 5 | 1844 | 56 | ||
XM_021242649.1 | XP_021098308.1 | 4 - 5 | 1900 | 56 | ||
XM_021242680.1 | XP_021098339.1 | 3 - 4 | 1718 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() |
