Assay Details
| Gene Symbol: LOC101698338 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004844514.2 | XP_004844571.1 | 12 - 13 | 1844 | 76 | |
| XM_004844515.2 | XP_004844572.1 | 12 - 13 | 1844 | 76 | ||
| XM_004844516.2 | XP_004844573.1 | 12 - 13 | 1844 | 76 | ||
More Information
Set Membership: |
|

LOC101698338
Genomic Map