Assay Details
Gene Symbol: Mmrn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004848078.2 | XP_004848135.1 | 3 - 4 | 1012 | 60 | |
XM_021261078.1 | XP_021116737.1 | 3 - 4 | 1012 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
