Assay Details
Gene Symbol: Rab11fip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004855378.3 | XP_004855435.1 | 2 - 3 | 820 | 76 | |
XM_004855380.3 | XP_004855437.1 | 2 - 3 | 920 | 76 | ||
XM_004855381.3 | XP_004855438.1 | 2 - 3 | 820 | 76 | ||
XM_021236822.1 | XP_021092481.1 | 2 - 3 | 544 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
