Assay Details
Gene Symbol: LOC101705115 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004862907.2 | XP_004862964.1 | 2 - 3 | 255 | 101 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004862907.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
