Assay Details
| Gene Symbol: Erich6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021257774.1 | XP_021113433.1 | 10 - 11 | 1729 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Erich6b| Gene Symbol: Erich6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021257774.1 | XP_021113433.1 | 10 - 11 | 1729 | 69 | |
Set Membership: |
|