Assay Details
Gene Symbol: Cnnm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004844375.2 | XP_004844432.1 | 4 - 5 | 1690 | 59 | |
XM_004844377.2 | XP_004844434.1 | 4 - 5 | 1690 | 59 | ||
XM_021258024.1 | XP_021113683.1 | 4 - 5 | 1690 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
