Assay Details
Gene Symbol: Aaed1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004846087.2 | XP_004846144.1 | 2 - 3 | 399 | 120 | |
XM_021259387.1 | XP_021115046.1 | 2 - 3 | 555 | 120 | ||
XM_021259388.1 | XP_021115047.1 | 2 - 3 | 576 | 120 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
