Assay Details
Gene Symbol: Aaed1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004846087.2 | XP_004846144.1 | 3 - 4 | 508 | 73 | |
XM_021259387.1 | XP_021115046.1 | 3 - 4 | 664 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
