Assay Details
| Gene Symbol: Ctla4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004851172.1 | XP_004851229.1 | 1 - 2 | 115 | 92 | |
| XM_004851173.1 | XP_004851230.1 | 1 - 2 | 115 | 92 | ||
More Information
Set Membership: |
|

Ctla4
Genomic Map