Assay Details
Gene Symbol: Lypd6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004869994.2 | XP_004870051.1 | 5 - 6 | 489 | 62 | |
XM_004869996.3 | XP_004870053.1 | 4 - 5 | 576 | 62 | ||
XM_004869997.3 | XP_004870054.1 | 5 - 6 | 705 | 62 | ||
XM_004869998.3 | XP_004870055.1 | 4 - 5 | 451 | 62 | ||
XM_004869999.3 | XP_004870056.1 | 4 - 5 | 477 | 62 | ||
XM_021248418.1 | XP_021104077.1 | 5 - 6 | 723 | 62 | ||
XM_021248419.1 | XP_021104078.1 | 5 - 6 | 660 | 62 | ||
XM_021248420.1 | XP_021104079.1 | 4 - 5 | 404 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
