Assay Details
Gene Symbol: Tmem79 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004871197.3 | XP_004871254.1 | 3 - 4 | 1157 | 59 | |
XM_021249534.1 | XP_021105193.1 | 3 - 4 | 993 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
