Assay Details
Gene Symbol: Cacng8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021245716.1 | XP_021101375.1 | 1 - 2 | 376 | 52 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
