Assay Details
Gene Symbol: Cacng8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021245716.1 | XP_021101375.1 | 2 - 3 | 461 | 110 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
