Assay Details
Gene Symbol: Cacng8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021245716.1 | XP_021101375.1 | 3 - 4 | 597 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
