Assay Details
| Gene Symbol: Shroom1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004836598.2 | XP_004836655.1 | 3 - 4 | 1146 | 76 | |
| XM_013078972.2 | XP_012934426.1 | 3 - 4 | 1092 | 76 | ||
| XM_021241411.1 | XP_021097070.1 | 3 - 4 | 1147 | 76 | ||
| XM_021241416.1 | XP_021097075.1 | 3 - 4 | 1647 | 76 | ||
More Information
Set Membership: |
|

Shroom1
Genomic Map