Assay Details
Gene Symbol: Lhfpl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004846699.2 | XP_004846756.1 | 1 - 2 | 684 | 79 | |
XM_021259939.1 | XP_021115598.1 | 1 - 2 | 684 | 79 | ||
XM_021259940.1 | XP_021115599.1 | 1 - 2 | 684 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
