Assay Details
Gene Symbol: Paip2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004849893.3 | XP_004849950.1 | 4 - 5 | 730 | 65 | |
XM_004849894.3 | XP_004849951.1 | 5 - 6 | 788 | 65 | ||
XM_013074086.2 | XP_012929540.1 | 5 - 6 | 755 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
