Assay Details
Gene Symbol: Lrrc56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004851721.3 | XP_004851778.2 | 9 - 10 | 2179 | 72 | |
XM_021264052.1 | XP_021119711.1 | 9 - 10 | 2422 | 72 | ||
XM_021264053.1 | XP_021119712.1 | 9 - 10 | 2419 | 72 | ||
XM_021264054.1 | XP_021119713.1 | 8 - 9 | 2392 | 72 | ||
XM_021264055.1 | XP_021119714.1 | 9 - 10 | 2386 | 72 | ||
XM_021264056.1 | XP_021119715.1 | 9 - 10 | 2422 | 72 | ||
XM_021264058.1 | XP_021119717.1 | 9 - 10 | 2277 | 72 | ||
XM_021264059.1 | XP_021119718.1 | 9 - 10 | 2280 | 72 | ||
XM_021264060.1 | XP_021119719.1 | 8 - 9 | 1973 | 72 | ||
XM_021264061.1 | XP_021119720.1 | 8 - 9 | 1096 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
