Assay Details
Gene Symbol: Dkk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004855446.2 | XP_004855503.1 | 3 - 4 | 669 | 84 | |
XM_021236747.1 | XP_021092406.1 | 4 - 5 | 6235 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() |
