Assay Details
Gene Symbol: Rabep2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004856035.3 | XP_004856092.1 | 7 - 8 | 1830 | 58 | |
XM_021237393.1 | XP_021093052.1 | 7 - 8 | 1877 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
