Assay Details
| Gene Symbol: Mtfr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004871489.2 | XP_004871546.1 | 7 - 8 | 3474 | 77 | |
| XM_021249569.1 | XP_021105228.1 | 7 - 8 | 3294 | 77 | ||
More Information
Set Membership: |
|

Mtfr2
Genomic Map