Assay Details
Gene Symbol: Rab43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004874855.1 | XP_004874912.1 | 2 - 3 | 519 | 62 | |
XM_013069301.2 | XP_012924755.1 | 2 - 3 | 516 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
