Assay Details
| Gene Symbol: LOC101719080 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021239264.1 | XP_021094923.1 | 1 - 2 | 388 | 77 | |
| XM_021239265.1 | XP_021094924.1 | 1 - 2 | 385 | 77 | ||
| XM_021239266.1 | XP_021094925.1 | 1 - 2 | 371 | 77 | ||
More Information
Set Membership: |
|

LOC101719080
Genomic Map