Assay Details
| Gene Symbol: LOC101719080 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021239264.1 | XP_021094923.1 | 2 - 3 | 567 | 102 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101719080| Gene Symbol: LOC101719080 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021239264.1 | XP_021094923.1 | 2 - 3 | 567 | 102 | |
Set Membership: |
|