Assay Details
Gene Symbol: LOC101726570 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004838016.3 | XP_004838073.1 | 1 - 2 | 317 | 107 | |
XM_004838018.3 | XP_004838075.1 | 1 - 2 | 348 | 107 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
