Assay Details
Gene Symbol: LOC101726570 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004838016.3 | XP_004838073.1 | 4 - 5 | 698 | 80 | |
XM_021252518.1 | XP_021108177.1 | 4 - 5 | 494 | 80 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
