Assay Details
Gene Symbol: Tsga13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004856319.2 | XP_004856376.1 | 6 - 7 | 752 | 75 | |
XM_021237589.1 | XP_021093248.1 | 7 - 8 | 788 | 75 | ||
XM_021237590.1 | XP_021093249.1 | 5 - 6 | 624 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |
