Assay Details
| Gene Symbol: Foxr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004856741.3 | XP_004856798.1 | 4 - 5 | 792 | 60 | |
| XM_021237739.1 | XP_021093398.1 | 4 - 5 | 709 | 60 | ||
| XM_021237740.1 | XP_021093399.1 | 4 - 5 | 709 | 60 | ||
| XM_021237741.1 | XP_021093400.1 | 5 - 6 | 767 | 60 | ||
| XM_021237742.1 | XP_021093401.1 | 4 - 5 | 709 | 60 | ||
More Information
Set Membership: |
|

Foxr1
Genomic Map